Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

α-Cyclodextrin ≥98%

5 kg
Catalog # SPCMC1620-5KGBL
Supplier:   Spectrum Chemical
α-Cyclodextrin ≥98%
Catalog # SPCMC1620-5KGBL
Supplier:  Spectrum Chemical
CAS Number:  10016-20-3
α-Cyclodextrin ≥98%

5 kg

CAS Number:  10016-20-3
Price
14 670,00 €
Log in for availability
1 * 5 kg
Product Details & Documents

alpha-Cyclodextrin, a polysaccharide, can be used as a dietary fiber.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Specifications

Specification Test Results

  • Assay (%):
    98

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.