Order Entry
Belgium
ContactUsLinkComponent
Iron(II) sulphate hydrate, dried powder FCC
Irritant
Iron(II) sulphate hydrate, dried powder FCC
Catalog # SPCMF1063-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Iron(II) sulphate hydrate, dried powder FCC
Catalog # SPCMF1063-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (FeSO₄):
    86.0 - 89.0%
  • Insoluble Substances:
    max. 0.05 %
  • Lead (Pb):
    max. 2 mg/kg
  • Mercury:
    max. 1 mg/kg
  • Identification:
    To Pass Test

Specification Test Results

About this item

 :
 :169.93 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:13463-43-9
Formula:FeSO₄·H₂O
MDL Number:MFCD00149722