Carboxymethyl cellulose sodium salt, 1500-3000 cP (1% H₂O - 25°C) USP, high viscosity
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:9004-32-4
:
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472
Storage Temperature:Ambient
Frequently Bought Together