Order Entry
Belgium
ContactUsLinkComponent
Sodium-L(+)-glutamate monohydrate ≥98.5% FCC
Sodium-L(+)-glutamate monohydrate ≥98.5% FCC
Supplier:  Spectrum Chemical
Certificates
Sodium-L(+)-glutamate monohydrate ≥98.5% FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • ASSAY (C5H8NNaO4; DRIED BASIS):
    98.5 - 101.5%
  • Optical Rotation:
    +24.5 to +25.5°
  • Clarity and Color of Solution:
    To Pass Test
  • Chloride (Cl):
    max. 0.2%
  • Lead (Pb):
    max. 5 mg/kg

Specification Test Results

Frequently Bought Together