Order Entry
Belgium
ContactUsLinkComponent
4-Aminophenol ≥98%
Irritant
Health
4-Aminophenol ≥98%
Catalog # SPCMA2305-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Health
4-Aminophenol ≥98%
Catalog # SPCMA2305-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

Specification Test Results

  • Assay:
    98% min.

Specification Test Results

About this item

 :
 :109.13 g/mol
Storage Temperature:Ambient
UN:2512
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:123-30-8
Formula:H₂NC₆H₄OH
MDL Number:MFCD00007869