Order Entry
Belgium
ContactUsLinkComponent
Trisodium citrate, anhydrous ≥99.0% USP
Trisodium citrate, anhydrous ≥99.0% USP
Supplier:  Spectrum Chemical
Certificates
Trisodium citrate, anhydrous ≥99.0% USP
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Anhydrous Basis):
    99.0 - 100.5%
  • Alkalinity:
    To Pass Test
  • Water Determination:
    max. 1.0 %
  • Tartrate:
    To Pass Test
  • Heavy Metals:
    max. 10 ppm

Specification Test Results

Frequently Bought Together