Order Entry
Belgium
ContactUsLinkComponent
Trisodium citrate, anhydrous ≥99.0%, powder FCC
Trisodium citrate, anhydrous ≥99.0%, powder FCC
Supplier:  Spectrum Chemical
Certificates
Trisodium citrate, anhydrous ≥99.0%, powder FCC
Supplier:  Spectrum Chemical
Certificates

Specification Test Results

  • Assay (C₆H₅Na₃O₇; Anhydrous Basis):
    99.0 - 100.5%
  • Lead:
    max. 2 mg/kg
  • Alkalinity:
    To Pass Test
  • Water Determination:
    max. 1 %
  • Identification:
    To Pass Test

Specification Test Results

Related Information

Frequently Bought Together