Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide solution 20% (w/v)
Sodium hydroxide solution 20% (w/v)
Catalog # SPCMS-229-500MLPL
Supplier:  Spectrum Chemical
CAS Number:  
Sodium hydroxide solution 20% (w/v)
Catalog # SPCMS-229-500MLPL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
UN:1824
ADR:8,II
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548