


1-Butanol NF
:71-36-3
:
:H₃C(CH₂)₂CH₂OH
:MFCD00002964
:74.12 g/mol
:13,01539
:117.6 °C (1013 hPa)
:–89 °C
:0,811
:30 °C
:Ambient
:200-751-6
:1120
:3,III
:01-2119484630-38
:GHS02
:GHS05
:GHS07
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Tubings, Original Perfusor®
: B. Braun