Order Entry
Belgium
ContactUsLinkComponent
Anti-C23 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-C23 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-8536R-A680
Supplier:  Bioss
Anti-C23 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-8536R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C23
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Epitope:
    601-710/710
  • Form:
    Liquid
  • Gene ID:
    4691
  • Antigen synonyms:
    Protein C23|Nucleolin|C23|NCL
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    76 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Nucleolin
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Nucleolin is the major nucleolar protein of growing eukaryotic cells. It is found associated with intranucleolar chromatin and pre-ribosomal particles. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly. May play a role in the process of transcriptional elongation. Binds RNA oligonucleotides with 5'-UUAGGG-3' repeats more tightly than the telomeric single-stranded DNA 5'-TTAGGG-3' repeats.

Type: Primary
Antigen: C23
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-710/710
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat