Specifications
- Antibody type:Primary
- Antigen name:LGI4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-200/537
- Form:Liquid
- Gene ID:163175
- Antigen synonyms:LGI1 like protein 3|Leucine rich glioma inactivated protein 4|LGI4|leucine-rich repeat LGI family member 4|Lgi4 protein|Leucine rich repeat LGI family member 4|LGI4_HUMAN|Lgil3|Leucine rich glioma inactivated gene 4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human LGI4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
LGI4, also known as leucine-rich glioma-inactivated protein 4, is a 537 amino acid secreted glycosylated protein that is widely expressed, with highest levels found within the nervous system. Interestingly, siRNA knockdown studies of LGI4 expression in Schwann cells have been shown to result in the inhibition of myelination, thus suggesting that LGI4 is an essential component of myelin formation and axon segregation. LGI4 shares significant homology with its other family members, LGI1, LGI2 and LGI3. Significantly, mutations in the gene encoding LGI1 have been linked to human temporal lobe epilepsy and, given the sequence similarity of LGI4, it is likely that it also may be implicated in the pathology of seizures. LGI4 is localised subcellularly to the Golgi, ER and vesicles. There are two isoforms of LGI4 that are produced as a result of alternative splicing events.
Type: Primary
Antigen: LGI4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/537
Host: Rabbit
Isotype: IgG
Reactivity: