Order Entry
Belgium
ContactUsLinkComponent
Anti-COLEC11 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-COLEC11 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-8032R-A750
Supplier:  Bioss
Anti-COLEC11 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-8032R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    COLEC11
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    122-200/271
  • Form:
    Liquid
  • Gene ID:
    78989
  • Antigen synonyms:
    Collectin 11|Collectin11|Collectin kidney I|CL K1 I|Collectin kidney protein 1|CL K1 IIb|MGC129471|DKFZp686N1868|MGC3279|CL K1 IIa|COLEC 11|Collectin sub family member 11|CL K1|COL11_HUMAN|CLK1|CL K1 II|MGC129470
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    26 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human COLEC11(kidney)
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

COLEC11 is a 271 amino acid C-type lectin protein that contains a collagen-like domain and a carbohydrate recognition domain, and plays an important role in host-defense. COLEC11 binds to various sugars and LPS (lipopolysaccharides), which include fucose but does not bind to glucose, hnRNP, Beta-1,3-Gal-T3 or mannose. COLEC11 is ubiquitously expressed in most tissues with high expression in kidney, liver, fetal liver, small intestine, thymus, spinal cord, placenta, adrenal gland, pancreas and several cell lines. COLEC11 is a secreted protein and all alternatively spliced isoforms of COLEC11 have oligomeric structures created through disulfide bonding.

Type: Primary
Antigen: COLEC11
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 122-200/271
Host: Rabbit
Isotype: IgG
Reactivity: