Order Entry
Belgium
ContactUsLinkComponent
 
Anti-RNA polymerase 2 CTD repeat YSPTSPS phospho Ser5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6582R-A680
 :  Bioss
Anti-RNA polymerase 2 CTD repeat YSPTSPS phospho Ser5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6582R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    RNA polymerase 2 CTD repeat YSPTSPS phospho Ser5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    6193
  • Antigen synonyms:
    DNA directed RNA polymerase II A|RNA polymerase II subunit B1|Polr2a|RNA pol II CTD|p-Rpb1 CTD Ser2/Ser5|DNA-directed RNA polymerase III largest subunit|RPB1|RNA-directed RNA polymerase II subunit RPB1|DNA-directed RNA polymerase II largest subunit|DNA-directed RNA polymerase II subunit RPB1|p-Rpb1 CTDSer2/Ser5|RPB1_HUMAN|DNA-directed RNA polymerase II subunit A
  • Modification:
    Phosphorylation (Ser5)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    240 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human RNA polymerase II CTD repeat YSPTSPS around the phosphorylation site of phospho S5.
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Largest and catalytic component of RNA polymerase II which synthesises mRNA precursors and many functional non-coding RNAs. Forms the polymerase active center together with the second largest subunit. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB1 is part of the core element with the central large cleft, the clamp element that moves to open and close the cleft and the jaws that are thought to grab the incoming DNA template. At the start of transcription, a single stranded DNA template strand of the promoter is positioned within the central active site cleft of Pol II. A bridging helix emanates from RPB1 and crosses the cleft near the catalytic site and is thought to promote translocation of Pol II by acting as a ratchet that moves the RNA-DNA hybrid through the active site by switching from straight to bent conformations at each step of nucleotide addition. During transcription elongation, Pol II moves on the template as the transcript elongates. Elongation is influenced by the phosphorylation status of the C-terminal domain (CTD) of Pol II largest subunit (RPB1), which serves as a platform for assembly of factors that regulate transcription initiation, elongation, termination and mRNA processing. Acts as a RNA-dependent RNA polymerase when associated with small delta antigen of Hepatitis delta virus, acting both as a replicate and transcriptase for the viral RNA circular genome.

Type: Primary
Antigen: RNA polymerase 2 CTD repeat YSPTSPS phospho Ser5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human