Anti-APOB48R Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-6336R-A750
: Bioss
- Antibody type:Primary
- Antigen name:APOB48R
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:1001-1088/1088
- Form:Liquid
- Gene ID:55911
- Antigen synonyms:apoB100R|Apolipoprotein B-100 receptor|Apolipoprotein B48 receptor|Apolipoprotein B100 receptor|apo b-100|apoB-100R|apoB-48R|APOB
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:115 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human APOB48R
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
APOB48R is a macrophage receptor that binds to the apolipoprotein B48 of dietary triglyceride (TG) rich lipoproteins. It may provide essential lipids, lipid soluble vitamins and other nutrients to reticuloendothelial cells.
Type: Primary
Antigen: APOB48R
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1001-1088/1088
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Others