Order Entry
Belgium
ContactUsLinkComponent
Anti-Lysyl tRNA synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-Lysyl tRNA synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6127R-A680
Supplier:  Bioss
Anti-Lysyl tRNA synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6127R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lysyl tRNA synthetase
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    301-400/482
  • Form:
    Liquid
  • Gene ID:
    6850
  • Antigen synonyms:
    KRS|Lysine--tRNA ligase|LysRS|SYK_HUMAN|Lysyl-tRNA synthetase|KARS2|Lysine tRNA ligase|KARS|KARS 2|KIAA0070
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    54 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human KARS2/Lysyl tRNA syntase
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Catalyzes the specific attachment of an amino acid to its cognate tRNA in a 2 step reaction: the amino acid (AA) is first activated by ATP to form AA-AMP and then transferred to the acceptor end of the tRNA. When secreted, acts as a signaling molecule that induces immune response through the activation of monocyte/macrophages. Catalyzes the synthesis of diadenosine oligophosphate (Ap4A), a signaling molecule involved in the activation of MITF transcriptional activity. Interacts with HIV-1 virus GAG protein, facilitating the selective packaging of tRNA(3)(Lys), the primer for reverse transcription initiation.

Type: Primary
Antigen: Lysyl tRNA synthetase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/482
Host: Rabbit
Isotype: IgG
Reactivity: