Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-5954R-A680
: Bioss
- Antibody type:Primary
- Antigen name:RASA3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:501-600/834
- Form:Liquid
- Gene ID:22821
- Antigen synonyms:GAPIII|RASA3|GAP1IP4BP|GAP1(IP4BP)|Ras GTPase-activating protein 3|Ins P4-binding protein
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:91 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RASA3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Inhibitory regulator of the Ras-cyclic AMP pathway. Binds inositol tetrakisphosphate (IP4) with high affinity. Might be a specific IP4 receptor.
Type: Primary
Antigen: RASA3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 501-600/834
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat