Order Entry
Belgium
ContactUsLinkComponent
Flt3/CD135 Lysate (Adult Normal)
Flt3/CD135 Lysate (Adult Normal)
Catalog # NOVUNBP2-11587
Supplier:  Novus Biologicals
CAS Number:  
undefined
Flt3/CD135 Lysate (Adult Normal)
Catalog # NOVUNBP2-11587
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    100 µG
  • Tested applications:
    WB

Specifications

About this item

Flt3/CD135 Lysate (Adult Normal)

Tested Applications: Western Blot.