Order Entry
Belgium
ContactUsLinkComponent
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11340R-A750
Supplier:  Bioss
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11340R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CPEB3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    251-350/698
  • Form:
    Liquid
  • Antigen synonyms:
    Cytoplasmic polyadenylation element binding protein 3|CPE-binding protein 3|CPEB 3|CPEB3_HUMAN|Cytoplasmic polyadenylation element-binding protein 3|hCPEB-3|CPE-BP3|KIAA0940|Cpeb3
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    76 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CPEB3
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The regulated translation of messenger RNA is essential for cell-cycle progression, establishment of the body plan during early development and modulation of key activities in the central nervous system. Cytoplasmic polyadenylation, one mechanism of controlling translation, is driven by cytoplasmic polyadenylation element binding proteins, called CPEBs. CPEB3 (cytoplasmic polyadenylation element binding protein 3) is a 698 amino acid protein that contains two RNA recognition motif (RRM) domains and, like other CPEB proteins, may play a role in the maturation of the central nervous system. CPEB3 exists as multiple alternatively spliced isoforms that are encoded by a gene which maps to human chromosome 10, which houses over 1200 genes and comprises nearly 4.5% of the human genome.

Type: Primary
Antigen: CPEB3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/698
Host: Rabbit
Isotype: IgG
Reactivity: