Order Entry
Belgium
ContactUsLinkComponent
Anti-MCM7/CDC47 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-MCM7/CDC47 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10319R-A680
Supplier:  Bioss
Anti-MCM7/CDC47 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10319R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MCM7/CDC47
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    1-100/719
  • Form:
    Liquid
  • Gene ID:
    4176
  • Antigen synonyms:
    MCM 2|P1.1 MCM3|PNAS 146|Mcm7|P85MCM|Homolog of S. cerevisiae Cdc47|MCM 7|PNAS146|CDC 47|MCM7_HUMAN|P1.1-MCM3|P1CDC47|CDC47 homolog|MCM2|DNA replication licensing factor MCM7|CDC47|Minichromosome maintenance complex component 7|Minichromosome maintenance deficient 7|Minichromosome Maintainence 7|CDABP0042|MCM7 minichromosome maintenance deficient 7|Minichromosome maintenance protein 7
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    81 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CDC47
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumour suppressor protein RB1/RB. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Type: Primary
Antigen: MCM7/CDC47
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/719
Host: Rabbit
Isotype: IgG
Reactivity: