Order Entry
Belgium
ContactUsLinkComponent
 
Anti-HINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-10228R-A680
 :  Bioss
Anti-HINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-10228R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    HINT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Environmentally Preferable:
  • Epitope:
    41-126/126
  • Form:
    Liquid
  • Antigen synonyms:
    histidine triad nucleotide binding protein 1|PKCI1|Protein kinase C inhibitor 1|Adenosine 5'-monophosphoramidase|PKCI 1|Protein kinase C-interacting protein 1|Adenosine 5' monophosphoramidase|HINT 1|Histidine triad nucleotide-binding protein 1|Protein kinase C interacting protein 1|HINT-1|HINT1_HUMAN|PRKCNH1|PKCI-1|histidine triad nucleotide binding protein|histidine triad nucleotide-binding protein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    14 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human HINT1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

The protein encoded by this gene can hydrolyse substrates such as AMP-morpholidate, AMP-N-alanine methyl ester, AMP-alpha-acetyl lysine methyl ester, and AMP-NH2. The encoded protein interacts with these substrates via a histidine triad motif, which is part of the loop that binds to the substrate. This gene has been found to be a tumour suppressing gene. Several transcript variants, but only one of them protein-coding, have been found for this gene.

Type: Primary
Antigen: HINT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-126/126
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse