Anti-Osterix Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-1110R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Osterix
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit,Human,Mouse
- Western blot:Yes
- Epitope:331-431/431
- Form:Liquid
- Gene ID:121340
- Antigen synonyms:SP7|osterix|OSX|Zinc finger protein osterix|Transcription factor Sp7|OI12|OI11
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Osterix
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Transcriptional activator essential for osteoblast differentiation. Binds to SP1 and EKLF consensus sequences and to other G/C-rich sequences (By similarity).
Type: Primary
Antigen: Osterix
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-431/431
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rabbit