Order Entry
Belgium
ContactUsLinkComponent
 
Anti-G3BP1Ser232 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-5376R-A680
 :  Bioss
Anti-G3BP1Ser232 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-5376R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    G3BP1Ser232
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    10146
  • Antigen synonyms:
    G3BP|GAP SH3 domain-binding protein 1|Ras GTPase-activating protein-binding protein 1|G3BP-1|ATP-dependent DNA helicase VIII|HDH-VIII|hDH VIII|G3BP1
  • Modification:
    Phosphorylation (Ser232)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    52-60 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human G3BP1 around the phosphorylation site of Ser232
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves MYC mRNA preferentially at the 3'-UTR. ATP- and magnesium-dependent helicase. Unwinds preferentially partial DNA and RNA duplexes having a 17 bp annealed portion and either a hanging 3' tail or hanging tails at both 5'- and 3'-ends. Unwinds DNA/DNA, RNA/DNA, and RNA/RNA substrates with comparable efficiency. Acts unidirectionally by moving in the 5' to 3' direction along the bound single-stranded DNA.

Type: Primary
Antigen: G3BP1Ser232
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human