Order Entry
Belgium
ContactUsLinkComponent
Anti-LONP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-LONP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4245R-A680
Supplier:  Bioss
Anti-LONP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4245R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    LONP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    361-460/959
  • Form:
    Liquid
  • Gene ID:
    9361
  • Antigen synonyms:
    Mitochondrial ATP-dependent protease Lon|Lon protease homolog, mitochondrial|hLON|LONP|Lon protease-like protein|PRSS15|LON|Serine protease 15|LonHS|LONP1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    98 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human LONP1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

ATP-dependent serine protease that mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides as well as certain short-lived regulatory proteins in the mitochondrial matrix. May also have a chaperone function in the assembly of inner membrane protein complexes. Participates in the regulation of mitochondrial gene expression and in the maintenance of the integrity of the mitochondrial genome. Binds to mitochondrial promoters and RNA in a single-stranded, site-specific, and strand-specific manner. May regulate mitochondrial DNA replication and/or gene expression using site-specific, single-stranded DNA binding to target the degradation of regulatory proteins binding to adjacent sites in mitochondrial promoters. Endogenous substrates include mitochondrial steroidogenic acute regulatory (StAR) protein.

Type: Primary
Antigen: LONP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 361-460/959
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat