Specifications
- Antibody type:Primary
- Antigen name:TGF beta 5/ TGF beta 1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Xenopus laevis
- Western blot:Yes
- Epitope:301-382/382
- Form:Liquid
- Gene ID:397778
- Antigen synonyms:Transforming growth factor beta 5|TGF-beta-1|TGF beta 5|Transforming growth factor beta-1.TGFB1_XENLA|Transforming Growth Factor 5|TGF-beta-5
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:12 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from Xenopus laevis TGF beta 5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Transforming Growth Factor beta 5 (TGF beta 5) is a member of the TGF beta family of growth factors. The TGF beta polypeptides are multifunctional; capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. Transformed, as well as nonneoplastic tissues, release transforming growth factors; and essentially all mammalian cells possess a specific TGF receptor. The multi-modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide.
Type: Primary
Antigen: TGF beta 5/ TGF beta 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-382/382
Host: Rabbit
Isotype: IgG
Reactivity: Xenopus laevis