Anti-KAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2812R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:KAT4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:501-600/1872
- Form:Liquid
- Gene ID:6872
- Antigen synonyms:TAF2A|Cell cycle G1 phase defect|TAF1 RNA polymerase II TATA box binding protein TBP associated factor 250kDa|Transcription initiation factor TFIID subunit 1|Transcription initiation factor TFIID 250 kDa subunit|BA2R|CCGS|NSCL2|NSCL 2|TAF II P250|TAFII-250|CCG1|TAF1 RNA polymerase II TATA box binding protein TBP associated factor|TBP Associated Factor 1|TAFII 250|OF|TBP associated factor 250 kDa|TAF1|Transcription factor TFIID p250 polypeptide|TAF1 RNA polymerase II TATA box binding protein associated factor 250kDa|TATA box binding protein TBP associated factor RNA polymerase II A 250kD|CCG 1|TAF 2A|TBP-associated factor 250 kDa|Cell cycle gene 1 protein|TAFII250|Complementation of cell cycle block G1 to S|p250|DYT3 protein|TATA box binding protein TBP associated factor RNA polymerase II A|TAF1_HUMAN|TAF 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:200 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TBP Associated Factor 1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Largest component and core scaffold of the TFIID basal transcription factor complex. Contains novel N- and C-terminal Ser/Thr kinase domains which can autophosphorylate or transphosphorylate other transcription factors. Phosphorylates TP53 on 'Thr-55' which leads to MDM2-mediated degradation of TP53. Phosphorylates GTF2A1 and GTF2F1 on Ser residues. Possesses DNA-binding activity. Essential for progression of the G1 phase of the cell cycle.
Type: Primary
Antigen: KAT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/1872
Host: Rabbit
Isotype: IgG
Reactivity: