Anti-RGS5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2794R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RGS5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:81-181/181
- Form:Liquid
- Gene ID:19737
- Antigen synonyms:RGS5|Regulator of G-protein signaling 5|1110070A02Rik
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:22 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse RGS5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G(i)-alpha and G(o)-alpha, but not to G(s)-alpha.
Type: Primary
Antigen: RGS5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 81-181/181
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat