Order Entry
Belgium
ContactUsLinkComponent
Anti-HIC2 Rabbit Polyclonal Antibody (FITC)
Anti-HIC2 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15486R-FITC
Supplier:  Bioss
Anti-HIC2 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15486R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HIC2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    501-600/615
  • Form:
    Liquid
  • Gene ID:
    23119
  • Antigen synonyms:
    Hic3|HIC1 related gene on chromosome 22|KIAA1020|ZNF907|ZBTB30|HRG22|HIC2_HUMAN|HIC2|Hypermethylated in cancer 2 protein|Zinc finger and BTB domain-containing protein 30
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    66 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human HIC2
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of Zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. HIC-2 (hypermethylated in cancer 2) possesses Zinc finger motifs that are thought to be important for DNA-binding and also has a BTB/POZ domain at the N-terminus, which is thought to be important for protein-protein binding, as well as for the binding of transcription factors. HIC-2 is also known as Hic-3, HIC1-related gene on chromosome 22 or Zinc finger and BTB domain-containing protein 30, and is a 615 amino acid protein that is expressed as two isoforms produced by alternative splicing. HIC-2 is highly expressed in cerebellum and is localised to the nucleus in cells. HIC-2 contains a short amino acid sequence that is thought to interact with CtBP, a transcriptional repressor. The gene sequence associated with HIC-2 is thought to be a target for miRNAs (microRNAs) which are expressed in many cancers, suggesting that HIC-2 could possess tumor suppressor capabilities.

Type: Primary
Antigen: HIC2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 501-600/615
Host: Rabbit
Isotype: IgG
Reactivity: