Saisie de commande
Belgium
ContactUsLinkComponent
Anti-HEXO Rabbit Polyclonal Antibody (Cy3)
Anti-HEXO Rabbit Polyclonal Antibody (Cy3)
Catalog # BOSSBS-15470R-CY3
Supplier:  Bioss
Anti-HEXO Rabbit Polyclonal Antibody (Cy3)
Catalog # BOSSBS-15470R-CY3
Supplier:  Bioss

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    HEXO
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Cy3®
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Mouse
  • Épitope:
    51-150/349
  • Formulaire:
    Liquid
  • ID de gène:
    90459
  • Synonymes antigène:
    ERI1|Histone mRNA 3'-exonuclease 1|Histone mRNA 3'-end-specific exoribonuclease|Eri-1 homolog|3'-5' exoribonuclease 1|3'EXO|3'HEXO|HEXO|THEX1|Protein 3'hExo|3'-5' exonuclease ERI1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    40 kDa
  • Température de stockage:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Température de transport:
    4 °C
  • Immunogène:
    KLH conjugated synthetic peptide derived from human HEXO
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Type de conditionnement:
    Vial
  • Cdt:
    100 µl

Specifications

About this item

RNA exonuclease that binds to the 3'-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. A 2' and 3'-hydroxyl groups at the last nucleotide of the histone 3'-end is required for efficient degradation of RNA substrates. Also able to degrade the 3'-overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi). Requires for binding the 5'-ACCCA-3' sequence present in stem-loop structure. Able to bind other mRNAs. Required for 5,8S rRNA 3'-end processing. Also binds to 5,8S ribosomal RNA. Binds with high affinity to the stem-loop structure of replication-dependent histone pre-mRNAs.

Type: Primary
Antigen: HEXO
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 51-150/349
Host: Rabbit
Isotype: IgG
Reactivity: Mouse