Order Entry
Belgium
ContactUsLinkComponent
Anti-H2BFWT Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Anti-H2BFWT Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15401R-A680
Supplier:  Bioss
Anti-H2BFWT Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15401R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    H2BFWT
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    1-100/175
  • Form:
    Liquid
  • Gene ID:
    158983
  • Antigen synonyms:
    H2BWT_HUMAN|Histone H2B type W-T|H2BFWT|H2B histone family member W testis-specic
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    20 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human H2BFWT
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

H2BFWT (H2B histone family member W testis-specific) is a 175 amino acid nuclear membrane histone that belongs to the histone H2B family. In contrast to most H2B histones, H2BFWT does not contain the conserved C-terminal residue involved in monoubiquitination, but is structurally indistinguishable from conventional H2B histones. Also varying from other H2B histones, H2BFWT does not participate in the recruitment of chromosome condensation factors or in the assembly of mitotic chromsomes. Expressed in testis, H2BFWT is present in sperm cells and may be essential to telomere function.

Type: Primary
Antigen: H2BFWT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/175
Host: Rabbit
Isotype: IgG
Reactivity: