Order Entry
Belgium
ContactUsLinkComponent
Anti-C9ORF140 Rabbit Polyclonal Antibody (FITC)
Anti-C9ORF140 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15314R-FITC
Supplier:  Bioss
Anti-C9ORF140 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15314R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C9orf140
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Epitope:
    301-394/394
  • Form:
    Liquid
  • Gene ID:
    89958
  • Antigen synonyms:
    TS/MDEP|2010317E24Rik|Protein C9orf140|SAPCD2|SAPC2_HUMAN|Chromosome 9 open reading frame 140|Suppressor APC domain containing protein 2|p42,3|C9orf140|Tumor specicity and mitosis phase-dependent expression protein|Suppressor APC domain containing 2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C9ORF140
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

C9orf140 (chromosome 9 open reading frame 140), also known as TS/MDEP (tumor specificity and mitosis phase-dependent expression protein) or p42.3, is a 394 amino acid nuclear and cytoplasmic protein encoded by a gene that maps to human chromosome 9q34.3. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.

Type: Primary
Antigen: C9orf140
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 301-394/394
Host: Rabbit
Isotype: IgG
Reactivity: Rat