Order Entry
Belgium
ContactUsLinkComponent
SD OLLAS/DYKDDDDK Epitope Tag Kit
SD OLLAS/DYKDDDDK Epitope Tag Kit
Catalog # NOVUNBK1-19035
Supplier:  Novus Biologicals
CAS Number:  
undefined
SD OLLAS/DYKDDDDK Epitope Tag Kit
Catalog # NOVUNBK1-19035
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    2 Vial
  • Description:
    SD OLLAS/DYKDDDDK Epitope tag kit
  • Species reactivity:
    Non-species specific

Specifications

About this item

SD OLLAS/DYKDDDDK Epitope Tag Kit

Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Non-species specific.