Order Entry
Belgium
ContactUsLinkComponent
 
ND OLLAS/DYKDDDDK Epitope Tag Kit
Catalog # NOVUNBK1-19033
Supplier:  Novus Biologicals
CAS Number:  
undefined
ND OLLAS/DYKDDDDK Epitope Tag Kit
Catalog # NOVUNBK1-19033
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    2 Vial
  • Description:
    ND OLLAS/DYKDDDDK Epitope tag kit
  • Species reactivity:
    Non-species specific

Specifications

About this item

ND OLLAS/DYKDDDDK Epitope Tag Kit

Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Non-species specific.