Order Entry
Belgium
ContactUsLinkComponent
Chromata ChIP Histone H3 [ac Lys4] Kit
Chromata ChIP Histone H3 [ac Lys4] Kit
Catalog # NOVUNBP1-71710
Supplier:  Novus Biologicals
CAS Number:  
undefined
Chromata ChIP Histone H3 [ac Lys4] Kit
Catalog # NOVUNBP1-71710
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    25 Tests
  • Description:
    Chromata ChIP Histone H3 [ac Lys4] kit
  • ChIP:
    Yes
  • Antigen symbol:
    Chromata ChIP Histone H3
  • Product type:
    KIT

Specifications

About this item

Chromata ChIP Histone H3 [ac Lys4] Kit

Tested Applications: Chromatin Immunoprecipitation.