Order Entry
Belgium
ContactUsLinkComponent
Anti-TDP43 Mouse Monoclonal Antibody [clone: 6H6E12]
Anti-TDP43 Mouse Monoclonal Antibody [clone: 6H6E12]
Catalog # MSPP60019-2-IG
undefined
Anti-TDP43 Mouse Monoclonal Antibody [clone: 6H6E12]
Catalog # MSPP60019-2-IG

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TAR DNA binding Protein
  • Antigen symbol:
    TDP43
  • Clonality:
    Monoclonal
  • Clone:
    6H6E12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    ALS10|TDP 43|TARDBP|SIG-39854-100
  • Storage buffer:
    PBS with 0,1% sodium azide and 50% glycerol pH 7,3.
  • Shipping temperature:
    Store at –20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Caprylic acid/ammonium sulfate precipitation
  • Pk:
    150 µl

Specifications

About this item

Transactivation response (TAR) DNA-binding protein of 43 kDa (also known as TARDBP or TDP-43) was first isolated as a transcriptional inactivator binding to the TAR DNA element of the HIV-1 virus. Neumann et al. (2006) found that a hyperphosphorylated, ubiquitinated, and cleaved form of TARDBP, known as pathologic TDP-43, is the major component of the tau-negative and ubiquitin-positive inclusions that characterise amyotrophic lateral sclerosis (ALS) and the most common pathological subtype of frontotemporal lobar degeneration (FTLD-U). 60019-2-Ig is a mouse monoclonal antibody recognizing the cleavage product of 20-30 kDa in addition to the native and phosphorylated forms of TDP-43. Immunohistochemical analyses of TDP-43 using this antibody detect both normal diffuse nuclear staining and insoluble inclusions in pathologic tissues. The epitope of 60019-2-Ig has been determined to locate at residues 203-209 by Hiroshi Tsuji et al. (2012), which is involved in the formation of pathologic TDP-43 aggregation. Notably this antibody only recognizes human TDP-43 but not reacts with mouse or rat TDP-43.

Western Blot:K-562 Cells, 1:5000-1:50000; IF: MCF-7 Cells, 1:10-1:100; IP:K-562 Cells, 1:500-1:5000; FC: MCF-7 Cells, N/A; IHC: Human Brain(FTLD-U) Tissue, N/A

Type: Primary
Antigen: TDP43
Clonality: Monoclonal
Clone: 6H6E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human