Order Entry
Belgium
ContactUsLinkComponent
TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedicals
TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedicals
Catalog # ICNA092020026
Supplier:  MP Biomedicals
CAS Number:  
undefined
TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedicals
Catalog # ICNA092020026
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    20 mL
  • Description:
    Defined serum replacement, 50X concentrate
  • Storage temperature:
    4 °C

Specifications

About this item

This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.

  • A universal serum replacement that supports human and mammalian cells
  • Completely free of virus, mycoplasma and other infectious agents
  • Contains no growth factors, steroid hormones, extraneous proteins or phenol red
  • Protein origin - human; Drug Master on file with the FDA

TCH is a fortified, multipurpose serum replacement for long-term culturing of primarily human anchorage-dependent and suspension cells. Highly effective in the production of biologicals, as well as in culturing bacteria cell culture and other mammalian cells.