Order Entry
Belgium
ContactUsLinkComponent
Anti-TIA1 Rabbit Polyclonal Antibody
Anti-TIA1 Rabbit Polyclonal Antibody
Catalog # MSPP12133-2-AP
Anti-TIA1 Rabbit Polyclonal Antibody
Catalog # MSPP12133-2-AP

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TIA1 cytotoxic granule-associated RNA binding protein
  • Antigen symbol:
    TIA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    TIA-1|WDM
  • Storage buffer:
    PBS with 0,1% sodium azide and 50% glycerol pH 7,3
  • Shipping temperature:
    Store at ‒20 °C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Pk:
    150 µl

Specifications

About this item

TIA1, also named as p40-TIA-1, is involved in alternative pre-RNA splicing and regulation of mRNA translation by binding to AU-rich elements (AREs) located in mRNA 3' untranslated regions (3' UTRs). It possesses nucleolytic activity against cytotoxic lymphocyte target cells. TIA1 may be involved in apoptosis. Two isoforms of this protein exist - 41kDa and 42kDa.

Western Blot:Jurkat Cells, 1:200 to 1:2000

Type: Primary
Antigen: TIA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat