Order Entry
Belgium
ContactUsLinkComponent
QuickDetect™ Total Cholesterol (Mouse) ELISA Kit
QuickDetect™ Total Cholesterol (Mouse) ELISA Kit
  ABCAAB285242-96
 :  Abcam
 :  
undefined
QuickDetect™ Total Cholesterol (Mouse) ELISA Kit
  ABCAAB285242-96
 :  Abcam
 :  

 

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    QuickDetect™ Total cholesterol (Mouse) ELISA kit, Size; 100 assays

 

 

Rapid sandwich ELISA kit to quantify Total cholesterol in mouse serum, plasma, culture supernatnat.

  • Easy, convenient and time-saving method to measure the level of total Cholesterol in mouse serum, plasma, culture supernatants and other biological fluids
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results
  • Detection Range: 78 to 5000 pg/ml

Cholesterol, is an organic molecule, a sterol (or modified steroid), a type of lipid molecule, and is biosynthesised by all animal cells, in which it is an essential structural component of animal cell membranes that is required to maintain both membrane structural integrity and fluidity. Cholesterol enables animal cells to dispense with a cell wall, thereby allowing animal cells to change shape and animals to move. Total cholesterol is a measure of the total amount of cholesterol in blood, including low-density lipoprotein (LDL) cholesterol and high-density lipoprotein (HDL) cholesterol. BioVision’s QuickDetect™ total cholesterol ELISA kit is a sandwich ELISA assay for measuring the quantity of mouse total cholesterol in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.