Order Entry
Belgium
ContactUsLinkComponent
Alanine Aminotransferase 1 (GPT) (Mouse) ELISA Kit
Alanine Aminotransferase 1 (GPT) (Mouse) ELISA Kit
Catalog # ABCAAB285263-96
Supplier:  Abcam
CAS Number:  
undefined
Alanine Aminotransferase 1 (GPT) (Mouse) ELISA Kit
Catalog # ABCAAB285263-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Alanine Aminotransferase 1 (GPT)
  • Description:
    Alanine Aminotransferase 1 (GPT) (Mouse) ELISA Kit
  • Tests per kit:
    100 assays

Specifications

About this item

A Sandwich ELISA kit to quantify Mouse GPT in serum, plasma and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of GPT in mouse serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range:1,56 - 100 mIU/ml
  • Sensitivity: < 0,938 mIU/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%

Alanine aminotransferase 1 catalyzes the reversible transamination between alanine and 2-oxoglutarate to form pyruvate and glutamate. Participates in cellular nitrogen metabolism and also in liver gluconeogenesis starting with precursors transported from skeletal muscles. BioVision’s GPT ELISA kit is a sandwich ELISA assay for the quantitative measurement of mouse GPT in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of mouse GPT captured from the samples.