Order Entry
Belgium
ContactUsLinkComponent
Mouse TIMP-1 PicoKine; ELISA Kit
Mouse TIMP-1 PicoKine; ELISA Kit
Catalog # BSBTEK0521
Supplier:  Boster Bio
CAS Number:  
undefined
Mouse TIMP-1 PicoKine; ELISA Kit
Catalog # BSBTEK0521
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    TIMP-1
  • Description:
    Mouse TIMP-1 PicoKine; ELISA kit, 96 wells/kit

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse TIMP-1.

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse TIMP1 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. TIMP1 tissue specificity: Found in fetal and adult tissues. Highest levels are found in bone. Also found in lung, ovary and uterus.

96wells/kit, with removable strips.