Order Entry
Belgium
ContactUsLinkComponent
Mouse BMP-4 PicoKine; ELISA Kit
Mouse BMP-4 PicoKine; ELISA Kit
Catalog # BSBTEK0316
Supplier:  Boster Bio
CAS Number:  
undefined
Mouse BMP-4 PicoKine; ELISA Kit
Catalog # BSBTEK0316
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse BMP-4 PicoKine; ELISA kit, 96 wells/kit

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse BMP-4.

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Bmp4 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell culture supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Bmp4 tissue specificity:

96wells/kit, with removable strips.