Order Entry
Belgium
ContactUsLinkComponent
 
Hemoglobin A1c (HbA1c) (Mouse) ELISA Kit
  ABCAAB285317-96
 :  Abcam
 :  
undefined
Hemoglobin A1c (HbA1c) (Mouse) ELISA Kit
  ABCAAB285317-96
 :  Abcam
 :  

 

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Hemoglobin A1c (HbA1c) (mouse) ELISA kit

 

 

A sandwich ELISA kit for the quantitative measurement of hemoglobin A1c in biological fluids.

  • Easy, convenient and time-saving method to measure the level of HbA1c in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 125 to 80 ng/ml
  • Sensitivity: <075 ng/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • No significant cross-reactivity or interference was observed

Glycated hemoglobin (HbA1c) is a form of hemoglobin that is measured primarily to identify average plasma glucose concentration. HbA1c is a measure of the beta-N-1-deoxy fructosyl component of hemoglobin. Glycated hemoglobin testing is recommended for both checking the blood sugar control in people who might be pre-diabetic and monitoring blood sugar control in patients with more elevated levels, termed diabetes mellitus. It provides far more revealing information on glycemic behavior than a fasting blood sugar value. BioVision's HbA1c ELISA kit is a sandwich ELISA assay for the quantitative measurement of HbA1c in mouse serum, plasma and cell culture supernatants. The density of color is proportional to the amount of HbA1c captured from the samples.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.