Order Entry
Belgium
ContactUsLinkComponent
PicoKine Mouse PD-L1/B7-H1; ELISA Kit
PicoKine Mouse PD-L1/B7-H1; ELISA Kit
Catalog # BSBTEK1450
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Mouse PD-L1/B7-H1; ELISA Kit
Catalog # BSBTEK1450
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse PD-L1/B7-H1 PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of mouse PD-L1/B7-H1.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect Mouse Pd L1/Cd274 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Pd L1/Cd274 tissue specificity: Highly expressed in the heart, thymus, skeletal muscle, and lung. Weakly expressed in the kidney, spleen, thyroid, and liver. Expressed on activated dendritic cells, B- cells and macrophages. Expressed in numerous tumor cells lines of lymphoid origin. .

96wells/kit, with removable strips.