Order Entry
Belgium
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

Supplier:   Tonbo Biosciences
Propidium Iodide
Supplier:  Tonbo Biosciences
Certificates
Propidium Iodide
Propidium Iodide

2 Options Available Below

Product Details & Documents

Propidium Iodide (PI) is a membrane-impermeant DNA binding dye that cannot penetrate viable cells. PI rapidly enters cells with compromised membranes and intercalates between base pairs allowing exclusion of non-viable cells from analysis of flow cytometry data. PI has a broad emission spectrum from 535-617 nm and can be detected in either FL2 or FL3 detectors. When used with Annexin V FITC, it is recommended to analyze PI in FL2.

  • Formulation: 10 mM NaH₂PO₄, 150 mM NaCl, 0,09% NaN₃, pH 7,2
Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 32,90 €
    32,90 €
    1 * 200 Tests
       32,90 €

  • 500 Tests
    Log in for availability
    Log in for availability
    1 * 500 Tests
    1 * 500 Tests - 65,80 €
    65,80 €
    1 * 500 Tests
       65,80 €