Order Entry
Belgium
ContactUsLinkComponent
QuickDetect™ Heparin (Human) ELISA Kit
QuickDetect™ Heparin (Human) ELISA Kit
Catalog # ABCAAB285267-96
Supplier:  Abcam
CAS Number:  
undefined
QuickDetect™ Heparin (Human) ELISA Kit
Catalog # ABCAAB285267-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Human
  • Description:
    QuickDetect™ Heparin (Human) ELISA kit
  • Tests per kit:
    100 assays

Specifications

About this item

A rapid sandwich ELISA kit for quantifying of Heparin in human serum and other biological fluid.

  • Easy, convenient and time-saving
  • Performed within 90 minutes
  • Detection Range: 9,3 - 300 ng/mL

Heparin, also known as unfractionated heparin (UFH), is medication which is used as an anticoagulant (blood thinner). Specifically it is used to treat and prevent deep vein thrombosis, pulmonary embolism, and arterial thromboembolism. It is also used in the treatment of heart attacks and unstable angina. It is given by injection into a vein. Other uses include inside test tubes and kidney dialysis machines.

BioVision's QuickDetectTM Heparin ELISA kit is a sandwich ELISA assay for measuring the quantity of human Heparin in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.

Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.