Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Corning® 100-Layer CellCube® Module, Corning
Corning® 100-Layer CellCube® Module, Corning
Supplier:   Corning
Corning® 100-Layer CellCube® Module, Corning
Supplier:  Corning
Certificates
Corning® 100-Layer CellCube® Module, Corning
2 Options Available Below
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 85 000cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Specifications for Product Family
Specifications
  • Catalog No:
  • CORN3032
  • CORN3244
  • Volume de milieu:
  • 6000 ml
  • 6000 ml
  • Surface:
  • Tissue culture treated
  • Tissue culture treated
Specifications
Product Family Options
Product Information
  • Désignation
    Surface de croissance
    Availability
    Price
  • 100-layer CellCube® module, polycarbonate/polystyrene, sterile
    85000 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 510,00 €
    3 510,00 €
    1 * 1 ST
       3 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    CORN3032
    Volume de milieu
    6000 ml
    Surface
    Tissue culture treated
  • 100-layer CellCube® module with silicone tubing and AseptiQuik™ G, polycarbonate/polystyrene, sterile
    85000 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 740,00 €
    2 740,00 €
    1 * 1 ST
       2 740,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    CORN3244
    Volume de milieu
    6000 ml
    Surface
    Tissue culture treated