Order Entry
Belgium
ContactUsLinkComponent
Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals
Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals
Catalog # ICNA091810454
Supplier:  MP Biomedicals
CAS Number:  
undefined
Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals
Catalog # ICNA091810454
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    1X Hanks' balanced salts (modified), without Mg and Ca, red

Specifications

About this item

Used to minimise cell attachment or aggregation.

  • Sterile
  • Suitable for cell culture
  • Formulation: Inorganic Salts: Potassium Chloride [KCl] 400.00000 mg/L, Potassium Phosphate Monobasic [KH2PO4] 60.00000 mg/L, Sodium Bicarbonate [NaHCO3] 350.00000 mg/L, Sodium Chloride [NaCl] 8000.00000 mg/L, Sodium Phosphate Dibasic [Na2HPO4] 47.50000 mg/L, Dextrose 1000.00000 mg/L, Phenol Red Sodium Salt 17.00000 mg/L
  • pH: 6,9-7,4
  • Osmolality 258 - 298 mM/kg

Calcium and magnesium support cell adhesion. Hanks' Balanced Salts (Modified), without Magnesium and Calcium, is used to minimise cell attachment or aggregation.