Order Entry
Belgium
ContactUsLinkComponent
PicoKine Human Tryptase/TPSAB1,B2; ELISA Kit
PicoKine Human Tryptase/TPSAB1,B2; ELISA Kit
Catalog # BSBTEK0898
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Human Tryptase/TPSAB1,B2; ELISA Kit
Catalog # BSBTEK0898
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Human Tryptase/TPSAB1,B2 PicoKine; ELISA kit
  • Tests per kit:
    96 wells/kit

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of human tryptase/TPSAB1,B2.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect human tryptase beta-2/TPSB2 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tryptase Beta-2/TPSB2 tissue specificity: TPSAB1: Isoform 1 and isoform 2 are expressed in lung,stomach, spleen, heart and skin; in these tissues, isoform 1 ispredominant. Isoform 2 is expressed in aorta, spleen, and breasttumor, with highest levels in the endothelial cells of some bloodvessels surrounding the aorta, as well as those surrounding thetumor and low levels, if any, in mast cells (at protein level).

96wells/kit, with removable strips.