Monocyte Attachment Medium
Catalog # MSPPC-28051
Supplier: MARKET SOURCE PART PROCESS
Specifications
- Pk:250 mL
Specifications
About this item
The Monocyte Attachment Medium allows for the efficient adherence selection of Monocytes from freshly isolated Mononuclear Cells (MNC) while maintaining optimal cell health.
Recommended for Human Mononuclear Cells, freshly isolated.
Thus, the time-consuming and costly immunomagnetic purification of monocytes can be skipped. With properly performed washing steps (see Instruction Manual) large numbers of 80 to 90% pure monocytes can be obtained with the PromoCell Monocyte Attachment Medium. Typical applications are generation of dendritic cells/macrophages as well as phagocytosis assays.
Ordering information: The Medium is ready-to-use and does not need further supplementation.