Order Entry
Belgium
ContactUsLinkComponent
Transthyretin (TTR) (Mouse) ELISA Kit
Transthyretin (TTR) (Mouse) ELISA Kit
Catalog # ABCAAB285262-96
Supplier:  Abcam
CAS Number:  
undefined
Transthyretin (TTR) (Mouse) ELISA Kit
Catalog # ABCAAB285262-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Mouse
  • Description:
    Transthyretin (TTR) (mouse) ELISA kit

Specifications

About this item

A Sandwich ELISA kit for the quantitative measurement of Mouse TTR in biological fluids.

  • Easy, convenient and time-saving method to measure the level of TTR in Mouse serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 7,81 to 500 ng/ml
  • Sensitivity: <4,68 ng/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%

Transthyretin (TTR) is also known as Prealbumin, ATTR, TBPA, PALB, which belongs to the transthyretin family. Transthyretin/TTR is a serum and cerebrospinal fluid carrier of the thyroid hormone thyroxine (T4) and retinol-binding protein bound to retinol. In cerebrospinal fluid TTR is the primary carrier of T4. TTR also acts as a carrier of retinol (vitamin A) through its association with etinol-binding protein (RBP) in the blood and the CSF. Less than 1% of TTR's T4 binding sites are occupied in blood. TTR misfolding and aggregation is known to be associated with the amyloid diseases.

BioVision’s Transthyretin ELISA kit is a sandwich ELISA assay for the quantitative measurement of human Transthyretin in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human Transthyretin captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.