Saisie de commande
Belgium
ContactUsLinkComponent
Reusable Respirator Spectacle Kit
Reusable Respirator Spectacle Kit
Catalog # MMMASK102
Supplier:  3M
undefined
Reusable Respirator Spectacle Kit
Catalog # MMMASK102
Supplier:  3M
Specifications
  • Désignation:
    Reusable respirator spectacle kit
  • Taille:
    Universal
  • Cdt:
    1 KIT
Specifications
About this item
Wear prescription lenses while using certain 3M™ Reusable Full Face Masks with the help of 3M™ Reusable Respirator Spectacle Kit. These spectacle frames attach to 3M™ Reusable Full Face Masks and can be adjusted.
  • Spectacle kit for 3M™ Reusable Full Face Masks
  • Permits wearer to use corrective lenses and to adjust them to a comfortable position
  • One each per case
  • Easy installation
  • For occupational use only
Get the correct level of comfort and protection from protective equipment, while using prescription spectacles, with the 3M™ Reusable Respirator Spectacle kit. The kit allows use of corrective lenses and adjusts their position for a suitable fit inside 3M™ Full Face Respirators. Easy to install, the spectacle kit helps to ensure worker safety and compliance for those workers requiring prescription eyewear.